Tisk…

Anti-TACC3 Mouse Monoclonal Antibody [clone: 8F6]

Dodavatel: OriGene
ORIGCF501253EA 21120 CZK
ORIGCF501253
Anti-TACC3 Mouse Monoclonal Antibody [clone: 8F6]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TACC3
Clonality: Monoclonal
Clone: 8F6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR