Tisk…

Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 3C8]

Dodavatel: OriGene
ORIGCF501274EA 21120 CZK
ORIGCF501274
Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 3C8]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: IGF2BP2
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR