Tisk…

Anti-GNMT Mouse Monoclonal Antibody [clone: 8A3]

Dodavatel: OriGene
ORIGCF501276EA 21710 CZK
ORIGCF501276
Anti-GNMT Mouse Monoclonal Antibody [clone: 8A3]
Protilátky
Recommended Dilutions: Western Blot: 1:500 Flow Cytometry: 1:100

Type: Primary
Antigen: GNMT
Clonality: Monoclonal
Clone: 8A3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG3
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR