Tisk…

Anti-ID2 Mouse Monoclonal Antibody [clone: 10C3]

Dodavatel: OriGene
ORIGCF500183EA 21120 CZK
ORIGCF500183
Anti-ID2 Mouse Monoclonal Antibody [clone: 10C3]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:50

Type: Primary
Antigen: ID2
Clonality: Monoclonal
Clone: 10C3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR