Tisk…

Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 3G4]

Dodavatel: OriGene
ORIGCF500240EA 21120 CZK
ORIGCF500240
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 3G4]
Protilátky
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR