Tisk…

Anti-CDX2 Mouse Monoclonal Antibody [clone: 3G4]

Dodavatel: OriGene
ORIGCF500252EA 21120 CZK
ORIGCF500252
Anti-CDX2 Mouse Monoclonal Antibody [clone: 3G4]
Protilátky
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CDX2
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR