Tisk…

Anti-MSI1 Mouse Monoclonal Antibody [clone: 2H9]

Dodavatel: OriGene
ORIGCF502253EA 21120 CZK
ORIGCF502253
Anti-MSI1 Mouse Monoclonal Antibody [clone: 2H9]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MSI1
Clonality: Monoclonal
Clone: 2H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR