Tisk…

Anti-DNAJB2 Mouse Monoclonal Antibody [clone: 5E5]

Dodavatel: OriGene
ORIGCF502263EA 21120 CZK
ORIGCF502263
Anti-DNAJB2 Mouse Monoclonal Antibody [clone: 5E5]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJB2
Clonality: Monoclonal
Clone: 5E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR