Tisk…

Anti-GPHN Mouse Monoclonal Antibody [clone: 7C6]

Dodavatel: OriGene
ORIGCF502283EA 21120 CZK
ORIGCF502283
Anti-GPHN Mouse Monoclonal Antibody [clone: 7C6]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: GPHN
Clonality: Monoclonal
Clone: 7C6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR