Tisk…

Anti-GBP5 Mouse Monoclonal Antibody [clone: 8D8]

Dodavatel: OriGene
ORIGCF502287EA 21710 CZK
ORIGCF502287
Anti-GBP5 Mouse Monoclonal Antibody [clone: 8D8]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP5
Clonality: Monoclonal
Clone: 8D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR