Tisk…

Anti-CTDSP1 Mouse Monoclonal Antibody [clone: 1F4]

Dodavatel: OriGene
ORIGCF502289EA 21120 CZK
ORIGCF502289
Anti-CTDSP1 Mouse Monoclonal Antibody [clone: 1F4]
Protilátky
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CTDSP1
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR