Tisk…

Anti-FOSL1 Mouse Monoclonal Antibody [clone: 12F9]

Dodavatel: OriGene
ORIGCF500624EA 21120 CZK
ORIGCF500624
Anti-FOSL1 Mouse Monoclonal Antibody [clone: 12F9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: FOSL1
Clonality: Monoclonal
Clone: 12F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR