Tisk…

Anti-GBP2 Mouse Monoclonal Antibody [clone: 1G1]

Dodavatel: OriGene
ORIGCF500658EA 21710 CZK
ORIGCF500658
Anti-GBP2 Mouse Monoclonal Antibody [clone: 1G1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP2
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR