Tisk…

Anti-USP5 Mouse Monoclonal Antibody [clone: 2F5]

Dodavatel: OriGene
ORIGCF501288EA 21710 CZK
ORIGCF501288
Anti-USP5 Mouse Monoclonal Antibody [clone: 2F5]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: USP5
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR