Tisk…

Anti-BDH2 Mouse Monoclonal Antibody [clone: 2G1]

Dodavatel: OriGene
ORIGCF501293EA 21120 CZK
ORIGCF501293
Anti-BDH2 Mouse Monoclonal Antibody [clone: 2G1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BDH2
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR