Tisk…

Anti-MPP3 Mouse Monoclonal Antibody [clone: 5C2]

Dodavatel: OriGene
ORIGCF501305EA 21710 CZK
ORIGCF501305
Anti-MPP3 Mouse Monoclonal Antibody [clone: 5C2]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MPP3
Clonality: Monoclonal
Clone: 5C2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR