Tisk…

Anti-HIBCH Mouse Monoclonal Antibody [clone: 2A5]

Dodavatel: OriGene
ORIGCF501318EA 21710 CZK
ORIGCF501318
Anti-HIBCH Mouse Monoclonal Antibody [clone: 2A5]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HIBCH
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR