Tisk…

Anti-SNX9 Mouse Monoclonal Antibody [clone: 2A6]

Dodavatel: OriGene
ORIGCF501322EA 21120 CZK
ORIGCF501322
Anti-SNX9 Mouse Monoclonal Antibody [clone: 2A6]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNX9
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR