Tisk…

Anti-SOX17 Mouse Monoclonal Antibody [clone: 2B6]

Dodavatel: OriGene
ORIGCF500281EA 21710 CZK
ORIGCF500281
Anti-SOX17 Mouse Monoclonal Antibody [clone: 2B6]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: SOX17
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR