Tisk…

Anti-SOX17 Mouse Monoclonal Antibody [clone: 5G6]

Dodavatel: OriGene
ORIGCF500282EA 21710 CZK
ORIGCF500282
Anti-SOX17 Mouse Monoclonal Antibody [clone: 5G6]
Protilátky
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SOX17
Clonality: Monoclonal
Clone: 5G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR