Tisk…

Anti-VEGF Mouse Monoclonal Antibody [clone: 2F7]

Dodavatel: OriGene
ORIGCF500288EA 21120 CZK
ORIGCF500288
Anti-VEGF Mouse Monoclonal Antibody [clone: 2F7]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: VEGF
Clonality: Monoclonal
Clone: 2F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR