Tisk…

Anti-USP13 Mouse Monoclonal Antibody [clone: 4G11]

Dodavatel: OriGene
ORIGCF500318EA 21120 CZK
ORIGCF500318
Anti-USP13 Mouse Monoclonal Antibody [clone: 4G11]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: USP13
Clonality: Monoclonal
Clone: 4G11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR