Tisk…

Anti-GFAP Mouse Monoclonal Antibody [clone: 4G8]

Dodavatel: OriGene
ORIGCF500337EA 21710 CZK
ORIGCF500337
Anti-GFAP Mouse Monoclonal Antibody [clone: 4G8]
Protilátky
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR