Tisk…

Anti-GFAP Mouse Monoclonal Antibody [clone: 4D11]

Dodavatel: OriGene
ORIGCF500338EA 21710 CZK
ORIGCF500338
Anti-GFAP Mouse Monoclonal Antibody [clone: 4D11]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR