Tisk…

Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 1H10]

Dodavatel: OriGene
ORIGCF502300EA 21120 CZK
ORIGCF502300
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 1H10]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 1H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR