Tisk…

Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 5D2]

Dodavatel: OriGene
ORIGCF502303EA 21120 CZK
ORIGCF502303
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 5D2]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 5D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR