Tisk…

Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 6E7]

Dodavatel: OriGene
ORIGCF502305EA 21120 CZK
ORIGCF502305
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 6E7]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 6E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR