Tisk…

Anti-PDE2A Mouse Monoclonal Antibody [clone: 5B10]

Dodavatel: OriGene
ORIGCF502319EA 21120 CZK
ORIGCF502319
Anti-PDE2A Mouse Monoclonal Antibody [clone: 5B10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDE2A
Clonality: Monoclonal
Clone: 5B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR