Tisk…

Anti-LTA4H Mouse Monoclonal Antibody [clone: 8F4]

Dodavatel: OriGene
ORIGCF500665EA 21710 CZK
ORIGCF500665
Anti-LTA4H Mouse Monoclonal Antibody [clone: 8F4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: LTA4H
Clonality: Monoclonal
Clone: 8F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR