Tisk…

Anti-RNF144B Mouse Monoclonal Antibody [clone: 7H2]

Dodavatel: OriGene
ORIGCF500698EA 21120 CZK
ORIGCF500698
Anti-RNF144B Mouse Monoclonal Antibody [clone: 7H2]
Protilátky
Recommended Dilutions: Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B
Clonality: Monoclonal
Clone: 7H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR