Tisk…

Anti-CD5 Mouse Monoclonal Antibody [clone: 4H3]

Dodavatel: OriGene
ORIGCF501333EA 21120 CZK
ORIGCF501333
Anti-CD5 Mouse Monoclonal Antibody [clone: 4H3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:100; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR