Tisk…

Anti-C21orf59 Mouse Monoclonal Antibody [clone: 1A1]

Dodavatel: OriGene
ORIGCF501360EA 21120 CZK
ORIGCF501360
Anti-C21orf59 Mouse Monoclonal Antibody [clone: 1A1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: C21orf59
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR