Tisk…

Anti-PROM2 Mouse Monoclonal Antibody [clone: 15F4]

Dodavatel: OriGene
ORIGCF500356EA 21710 CZK
ORIGCF500356
Anti-PROM2 Mouse Monoclonal Antibody [clone: 15F4]
Protilátky
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PROM2
Clonality: Monoclonal
Clone: 15F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR