Tisk…

Anti-USP13 Mouse Monoclonal Antibody [clone: 5B9]

Dodavatel: OriGene
ORIGCF500367EA 21120 CZK
ORIGCF500367
Anti-USP13 Mouse Monoclonal Antibody [clone: 5B9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: USP13
Clonality: Monoclonal
Clone: 5B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR