Tisk…

Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 5B12]

Dodavatel: OriGene
ORIGCF500371EA 21120 CZK
ORIGCF500371
Anti-SERPINA1 Mouse Monoclonal Antibody [clone: 5B12]
Protilátky
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SERPINA1
Clonality: Monoclonal
Clone: 5B12
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR