Tisk…

Anti-RDH14 Mouse Monoclonal Antibody [clone: 4G8]

Dodavatel: OriGene
ORIGCF502365EA 21120 CZK
ORIGCF502365
Anti-RDH14 Mouse Monoclonal Antibody [clone: 4G8]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000

Type: Primary
Antigen: RDH14
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR