Tisk…

Anti-PRL Mouse Monoclonal Antibody [clone: 6B1]

Dodavatel: OriGene
ORIGCF500719EA 21710 CZK
ORIGCF500719
Anti-PRL Mouse Monoclonal Antibody [clone: 6B1]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 6B1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR