Tisk…

Anti-CYP2E1 Mouse Monoclonal Antibody [clone: 5B9]

Dodavatel: OriGene
ORIGCF500734EA 21120 CZK
ORIGCF500734
Anti-CYP2E1 Mouse Monoclonal Antibody [clone: 5B9]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CYP2E1
Clonality: Monoclonal
Clone: 5B9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR