Tisk…

Anti-XPNPEP1 Mouse Monoclonal Antibody [clone: 1E3]

Dodavatel: OriGene
ORIGCF501383EA 21120 CZK
ORIGCF501383
Anti-XPNPEP1 Mouse Monoclonal Antibody [clone: 1E3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: XPNPEP1
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR