Tisk…

Anti-SSB Mouse Monoclonal Antibody [clone: 3H9]

Dodavatel: OriGene
ORIGCF500410EA 21120 CZK
ORIGCF500410
Anti-SSB Mouse Monoclonal Antibody [clone: 3H9]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SSB
Clonality: Monoclonal
Clone: 3H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR