Tisk…

Anti-ZFP36 Mouse Monoclonal Antibody [clone: 1G9]

Dodavatel: OriGene
ORIGCF502377EA 21120 CZK
ORIGCF502377
Anti-ZFP36 Mouse Monoclonal Antibody [clone: 1G9]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ZFP36
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR