Tisk…

Anti-ZFP36 Mouse Monoclonal Antibody [clone: 5E3]

Dodavatel: OriGene
ORIGCF502381EA 21120 CZK
ORIGCF502381
Anti-ZFP36 Mouse Monoclonal Antibody [clone: 5E3]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZFP36
Clonality: Monoclonal
Clone: 5E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR