Tisk…

Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 3F9]

Dodavatel: OriGene
ORIGCF502407EA 21120 CZK
ORIGCF502407
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 3F9]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR