Tisk…

Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 3C7]

Dodavatel: OriGene
ORIGCF500767EA 21120 CZK
ORIGCF500767
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 3C7]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: AKR1A1
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR