Tisk…

Anti-HSPA1A Mouse Monoclonal Antibody [clone: 5F3]

Dodavatel: OriGene
ORIGCF500770EA 21120 CZK
ORIGCF500770
Anti-HSPA1A Mouse Monoclonal Antibody [clone: 5F3]
Protilátky
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 5F3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR