Tisk…

Anti-TTLL12 Mouse Monoclonal Antibody [clone: 3H6]

Dodavatel: OriGene
ORIGCF500776EA 21710 CZK
ORIGCF500776
Anti-TTLL12 Mouse Monoclonal Antibody [clone: 3H6]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TTLL12
Clonality: Monoclonal
Clone: 3H6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR