Tisk…

Anti-PARP16 Mouse Monoclonal Antibody [clone: 4E2]

Dodavatel: OriGene
ORIGCF500453EA 21120 CZK
ORIGCF500453
Anti-PARP16 Mouse Monoclonal Antibody [clone: 4E2]
Protilátky
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: PARP16
Clonality: Monoclonal
Clone: 4E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR