Tisk…

Anti-SERPINC1 Mouse Monoclonal Antibody [clone: 8D5]

Dodavatel: OriGene
ORIGCF500455EA 21120 CZK
ORIGCF500455
Anti-SERPINC1 Mouse Monoclonal Antibody [clone: 8D5]
Protilátky
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINC1
Clonality: Monoclonal
Clone: 8D5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR