Tisk…

Anti-MAPK1 Mouse Monoclonal Antibody [clone: 1C11]

Dodavatel: OriGene
ORIGCF500474EA 21120 CZK
ORIGCF500474
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 1C11]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunoprecipitation: 2-4?g/mg

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 1C11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR