Tisk…

Anti-C21orf34 Mouse Monoclonal Antibody [clone: 5D1]

Dodavatel: OriGene
ORIGCF502428EA 21120 CZK
ORIGCF502428
Anti-C21orf34 Mouse Monoclonal Antibody [clone: 5D1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: C21orf34
Clonality: Monoclonal
Clone: 5D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR